Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BNC1
Entrez Gene ID
102168944
Gene Name
basonuclin 1
Gene Aliases
-
Chromosome Location
Chr.21: 24191922 - 24220409 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066593.1 XP_017922082.1 2 - 3 243 59

View Full Product Details