Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABHD8
Entrez Gene ID
102174923
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
Chromosome Location
Chr.7: 102957749 - 102964871 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682126.3 XP_005682183.2 2 - 3 955 61
GenBank mRNA JO424475.1 - 3 - 4 1159 61
KA347160.1 - 3 - 4 1113 61

View Full Product Details