Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNTF
Entrez Gene ID
102175228
Gene Name
ciliary neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.15: 1447796 - 1450492 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690293.3 XP_005690350.1 1 - 2 542 73

View Full Product Details