Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FOXR1
Entrez Gene ID
102171354
Gene Name
forkhead box R1
Gene Aliases
-
Chromosome Location
Chr.15: 52778790 - 52789043 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058992.1 XP_017914481.1 2 - 3 239 86

View Full Product Details