Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNK17
Entrez Gene ID
102185792
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
-
Chromosome Location
Chr.23: 35645363 - 35659153 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039322.1 XP_017894811.1 4 - 5 824 85

View Full Product Details