Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXO48
Entrez Gene ID
102183883
Gene Name
F-box protein 48
Gene Aliases
-
Chromosome Location
Chr.11: 66583198 - 66588194 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686820.3 XP_005686877.1 3 - 4 589 97
XM_005686821.3 XP_005686878.1 3 - 4 577 97

View Full Product Details