Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRP9
Entrez Gene ID
102188242
Gene Name
signal recognition particle 9
Gene Aliases
-
Chromosome Location
Chr.16: 27368876 - 27379483 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690525.3 XP_005690582.1 1 - 2 366 80

View Full Product Details