Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM52
Entrez Gene ID
102186982
Gene Name
transmembrane protein 52
Gene Aliases
-
Chromosome Location
Chr.16: 49316893 - 49318898 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060492.1 XP_017915981.1 4 - 5 372 57

View Full Product Details