Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMP28
Entrez Gene ID
102184088
Gene Name
matrix metallopeptidase 28
Gene Aliases
-
Chromosome Location
Chr.19: 14234173 - 14259498 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064218.1 XP_017919707.1 6 - 7 1237 74

View Full Product Details