Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FOXO6
Entrez Gene ID
102185425
Gene Name
forkhead box O6
Gene Aliases
-
Chromosome Location
Chr.3: 15982853 - 16004564 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013962879.2 XP_013818333.2 1 - 2 873 124
GenBank mRNA JO362071.2 - 1 - 2 464 124

View Full Product Details