Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RASD1
Entrez Gene ID
102171188
Gene Name
ras related dexamethasone induced 1
Gene Aliases
-
Chromosome Location
Chr.19: 34466189 - 34468559 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693625.3 XP_005693682.1 1 - 2 1053 112
GenBank mRNA JT711788.1 - 3 - 4 352 112

View Full Product Details