Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTF1A
Entrez Gene ID
102176975
Gene Name
pancreas associated transcription factor 1a
Gene Aliases
-
Chromosome Location
Chr.13: 23585624 - 23587516 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056879.1 XP_017912368.1 2 - 2 1069 99

View Full Product Details