Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FBXL8
Entrez Gene ID
102179975
Gene Name
F-box and leucine rich repeat protein 8
Gene Aliases
-
Chromosome Location
Chr.18: 36068279 - 36073595 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062108.1 XP_017917597.1 3 - 3 2788 95
GenBank mRNA JO434564.2 - 1 - 1 464 95
JT678377.1 - 7 - 7 1799 95

View Full Product Details