Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Timp2
Entrez Gene ID
100135629
Gene Name
TIMP metallopeptidase inhibitor 2
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176343: 3832975 - 3873130 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173024.1 NP_001166495.1 3 - 4 540 73
GenBank mRNA AF127803.1 - 3 - 4 540 73

View Full Product Details