Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lalba
Entrez Gene ID
100379577
Gene Name
lactalbumin alpha
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176410: 50981327 - 50983368 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172889.1 NP_001166360.1 2 - 3 375 100
GenBank mRNA J00051.1 - 2 - 3 375 100

View Full Product Details