Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serpinb6
Entrez Gene ID
100192323
Gene Name
serpin family B member 6
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176381: 299552 - 309841 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173045.1 NP_001166516.1 2 - 3 379 93
GenBank mRNA FJ234418.1 - 2 - 3 379 93

View Full Product Details