Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnj13
Entrez Gene ID
100379277
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176406: 29051438 - 29054646 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172754.1 NP_001166225.1 2 - 3 480 69
GenBank mRNA AF200713.1 - 2 - 3 480 69
AJ312242.1 - 1 - 2 52 69

View Full Product Details