Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mcpt3
Entrez Gene ID
100379541
Gene Name
mast cell protease 3
Gene Aliases
Mcp-3
Chromosome Location
Chr.Un|NT_176387: 12656558 - 12658470 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172838.2 NP_001166309.1 2 - 3 248 96
GenBank mRNA AB014340.1 - 2 - 3 249 96

View Full Product Details