Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ftl
Entrez Gene ID
100379558
Gene Name
ferritin light chain
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176349: 8357697 - 8359001 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172858.1 NP_001166329.1 1 - 2 219 63
GenBank mRNA AF230928.1 - 1 - 2 219 63
AF233445.1 - 1 - 2 219 63
EB369836.1 - 1 - 2 225 63

Gene Symbol
LOC100717203
Entrez Gene ID
100717203
Gene Name
ferritin light chain
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176401: 26161047 - 26161953 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005005057.2 XP_005005114.1 1 - 1 311 63

View Full Product Details