Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ftl
Entrez Gene ID
100379558
Gene Name
ferritin light chain
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176349: 8357697 - 8359001 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172858.1 NP_001166329.1 3 - 4 497 89
GenBank mRNA AF230928.1 - 3 - 4 497 89
AF233445.1 - 3 - 4 89
EB369836.1 - 3 - 4 89

View Full Product Details