Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kek4
Entrez Gene ID
34718
Gene Name
kekkon 4
Gene Aliases
CG9431, CT26742, Dmel\CG9431, Dmel_CG9431, kek-4
Chromosome Location
Chr.2L: 13212335 - 13215350 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032484
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273506.1 NP_001260435.1 2 - 3 2051 62
NM_001273507.1 NP_001260436.1 2 - 3 1874 62
NM_135771.2 NP_609615.2 2 - 3 1895 62
GenBank mRNA AF507920.1 - 1 - 2 1670 62

View Full Product Details