Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp28a5
- Entrez Gene ID
-
34817
- Gene Name
- CG8864-PA
- Gene Aliases
-
28a5, BEST:GH10483, BG:DS00180.11, CG8864, CYP28A5, CYP28a5, Cyp28A5, DS00180.11, Dmel\CG8864, Dmel_CG8864, cyp28a5, gh10483
- Chromosome Location
-
Chr.2L: 13977302 - 13979569 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028940
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135850.3 |
NP_609694.1 |
1 - 2 |
252 |
87 |
GenBank mRNA |
AY051533.1 |
- |
1 - 2 |
228 |
87 |
View Full Product Details