Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slif
Entrez Gene ID
40510
Gene Name
slimfast
Gene Aliases
CG11128, Dmel\CG11128, Dmel_CG11128, SLIF, Slif
Chromosome Location
Chr.3L: 22881045 - 22893821 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0037203
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_141171.3 NP_649428.1 2 - 3 1391 65
NM_168977.2 NP_730764.1 2 - 3 1482 65
NM_168978.2 NP_730765.1 2 - 3 1261 65
GenBank mRNA AY094824.1 - 2 - 3 1333 65

View Full Product Details