Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nompA
Entrez Gene ID
45587
Gene Name
no mechanoreceptor potential A
Gene Aliases
CG13207, Dmel\CG13207, Dmel_CG13207, NOMPA, Nomp-A, NompA, nomp, y
Chromosome Location
Chr.2R: 11310335 - 11318676 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016047
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299383.1 NP_001286312.1 6 - 7 1229 81
NM_080092.4 NP_524831.2 6 - 7 1229 81
NM_165821.4 NP_725030.2 6 - 7 1229 81
NM_165822.4 NP_725031.2 5 - 6 1100 81
NM_206096.3 NP_995818.1 5 - 6 1100 81

View Full Product Details