Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Galphaq
- Entrez Gene ID
-
36384
- Gene Name
- G protein alpha q subunit
- Gene Aliases
-
CG17759, DGalpha[[q]], DGalphaq, DGq, DGqalpha, Dgq, DmGalpha[[q]], Dmel\CG17759, Dmel_CG17759, Dromel_CG17759_galpha49b_mORF, G alpha 49B, G?q, GNAQ/11/14, G[[a]]q, G[[alphaq]], G[[q]], G[[q]]alpha, G[[qalpha]], G[q], Galpha, Galpha49B, Galpha49b, Galpha[[q]], Gaq, Gq, Gqa-3, Gqalpha, Gqalpha[[e]], Gqalphae, Gqalpha{e}, dG[[alphaq]], dG[[q]]alpha, dGaalpha, dGq, dGqa-49b, dGqalpha, dGqalpha-3, dGqalpha-49b, dgq, dgqalpha, galpha49B, sast
- Chromosome Location
-
Chr.2R: 12612740 - 12623197 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004435
- Gene Symbol
- CG45086
- Entrez Gene ID
-
19835902
- Gene Name
- CG45086-PA
- Gene Aliases
-
Dmel\CG45086, Dmel_CG45086, Dromel_CG17759_galpha49b_uORF
- Chromosome Location
-
Chr.2R: 12612740 - 12619820 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266487
View Full Product Details