Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
muskelin
Entrez Gene ID
36386
Gene Name
CG8811-PA
Gene Aliases
CG8811, DmMK, Dmel\CG8811, Dmel_CG8811
Chromosome Location
Chr.2R: 12628697 - 12632378 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033757
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273992.1 NP_001260921.1 3 - 4 876 70
NM_136957.3 NP_610801.1 3 - 4 876 70
NM_165928.1 NP_725200.1 4 - 5 910 70
GenBank mRNA BT150328.1 - 4 - 5 909 70

View Full Product Details