Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Csas
- Entrez Gene ID
-
317923
- Gene Name
- CMP-sialic acid synthase
- Gene Aliases
-
CG32220, CSAS, DmCSAS, Dmel\CG32220, Dmel_CG32220
- Chromosome Location
-
Chr.3L: 19988027 - 19988991 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052220
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168828.3 |
NP_730474.2 |
2 - 3 |
277 |
78 |
View Full Product Details