Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sick
Entrez Gene ID
35277
Gene Name
sickie
Gene Aliases
CG10662, CG13958, CG13959, CG13960, CG13961, CG13964, CG33316, CG34343, CG42589, CG43720, Dm_2L:104545, Dmel\CG43720, Dmel_CG43720
Chromosome Location
Chr.2L: 19796365 - 19958424 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263873
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103713.4 NP_001097183.1 3 - 4 519 68
NM_001273674.1 NP_001260603.1 5 - 6 1382 68
NM_001273676.1 NP_001260605.1 3 - 4 635 68
NM_001273677.1 NP_001260606.1 4 - 5 2401 68
NM_001273678.1 NP_001260607.1 4 - 5 3087 68
NM_001273680.2 NP_001260609.1 5 - 6 1287 68
NM_001273681.1 NP_001260610.1 3 - 4 840 68
NM_206007.5 NP_995729.2 5 - 6 1287 68
NM_206008.4 NP_995730.2 5 - 6 3254 68

View Full Product Details