Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sick
Entrez Gene ID
35277
Gene Name
sickie
Gene Aliases
CG10662, CG13958, CG13959, CG13960, CG13961, CG13964, CG33316, CG34343, CG42589, CG43720, Dm_2L:104545, Dmel\CG43720, Dmel_CG43720
Chromosome Location
Chr.2L: 19796365 - 19958424 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263873
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103713.4 NP_001097183.1 8 - 9 3130 67
NM_001273674.1 NP_001260603.1 10 - 11 3993 67
NM_001273676.1 NP_001260605.1 8 - 9 3246 67
NM_001273677.1 NP_001260606.1 9 - 10 5012 67
NM_001273678.1 NP_001260607.1 9 - 10 5698 67
NM_001273681.1 NP_001260610.1 8 - 9 3451 67
NM_206007.5 NP_995729.2 10 - 11 3898 67
NM_206008.4 NP_995730.2 10 - 11 5865 67

View Full Product Details