Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vari
Entrez Gene ID
35343
Gene Name
varicose
Gene Aliases
38E.21, CG9326, Dmel\CG9326, Dmel_CG9326, Vari, l(2)03953, l(2)03953b, l(2)38EFa, szar
Chromosome Location
Chr.2L: 20788662 - 20797042 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0250785
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273716.1 NP_001260645.1 7 - 8 1545 60
NM_165342.4 NP_724288.3 8 - 9 1946 60
NM_165343.3 NP_724289.2 7 - 8 1486 60
NM_206011.3 NP_995733.2 8 - 9 1883 60
GenBank mRNA BT056231.1 - 8 - 9 1882 60
BT132965.1 - 8 - 9 1944 60

View Full Product Details