Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
qvr
Entrez Gene ID
2768718
Gene Name
quiver
Gene Aliases
BcDNA:GH06755, CG30032, CG33472, Dmel\CG33472, Dmel_CG33472, anon-WO02059370.1, mdcds_22455, sss
Chromosome Location
Chr.2R: 11442505 - 11451604 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260499
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144174.3 NP_001137646.1 5 - 6 882 60
NM_001299389.1 NP_001286318.1 5 - 6 873 60

View Full Product Details