Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mam
- Entrez Gene ID
-
36555
- Gene Name
- mastermind
- Gene Aliases
-
CG8118, DMam, Dm0064, Dmel\CG8118, Dmel_CG8118, MAM, Mam, MamH, N-2G, SR2-4, dMam, l(2)03505, l(2)04615, mam(CAG)[[7]]
- Chromosome Location
-
Chr.2R: 13991203 - 14060356 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002643
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206109.2 |
NP_995831.1 |
3 - 4 |
1826 |
125 |
GenBank mRNA |
BT044353.1 |
- |
3 - 4 |
1811 |
125 |
View Full Product Details