Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CheB53a
Entrez Gene ID
3346205
Gene Name
Chemosensory protein B 53a
Gene Aliases
CG16797, CG33457, CG33550, Dmel\CG33550, Dmel_CG33550
Chromosome Location
Chr.2R: 16466709 - 16467522 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261292
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014530.1 NP_001014530.1 1 - 2 237 96

View Full Product Details