Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab4
Entrez Gene ID
36992
Gene Name
CG4921-PA
Gene Aliases
CG 4921, CG4921, DRAB4, DmRab4, Dmel\CG4921, Dmel_CG4921, Q7KY04, RAB4c, rab4
Chromosome Location
Chr.2R: 17573462 - 17574979 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016701
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079053.4 NP_523777.1 3 - 4 237 70
NM_166240.3 NP_725696.1 2 - 3 336 70
GenBank mRNA AB035670.1 - 2 - 3 290 70

View Full Product Details