Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grip
Entrez Gene ID
50391
Gene Name
Glutamate receptor interacting protein
Gene Aliases
CG14447, CG5980, DGrip, Dgrip, Dmel\CG14447, Dmel_CG14447, GRIP, dGRIP, dgrip, grip
Chromosome Location
Chr.X: 5966279 - 5986699 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029830
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297991.1 NP_001284920.1 8 - 9 2543 80
NM_001297992.1 NP_001284921.1 7 - 8 2494 80
NM_132057.3 NP_572285.2 7 - 8 2681 80
GenBank mRNA BT044143.1 - 7 - 8 2680 80

View Full Product Details