Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btv
Entrez Gene ID
35073
Gene Name
beethoven
Gene Aliases
CG15148, DHC, DHC2, DHC36D, Dhc36D, Dmel\CG15148, Dmel_CG15148
Chromosome Location
Chr.2L: 17962625 - 17982472 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023096
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042904.4 NP_001036369.3 9 - 10 4112 76

View Full Product Details