Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACXB
Entrez Gene ID
53427
Gene Name
Adenylyl cyclase X B
Gene Aliases
AC, AC34A, Ac34A, CG17174, DACXB, Dmel\CG17174, Dmel_CG17174
Chromosome Location
Chr.2L: 12916814 - 12921008 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138989.3 NP_620474.2 11 - 12 2916 91

View Full Product Details