Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- COX7C
- Entrez Gene ID
-
50246
- Gene Name
- Cytochrome c oxidase subunit 7C
- Gene Aliases
-
BcDNA:LD14731, CG2249, COXO, CoAVIIc, CoVIIc, Dmel\CG2249, Dmel_CG2249, VIIc, anon-EST:fe3A1
- Chromosome Location
-
Chr.2R: 10048353 - 10050066 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040773
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY118515.1 |
- |
3 - 4 |
1081 |
72 |
- Gene Symbol
- CG44296
- Entrez Gene ID
-
19835750
- Gene Name
- CG44296-PA
- Gene Aliases
-
Dmel\CG44296, Dmel_CG44296
- Chromosome Location
-
Chr.2R: 10050348 - 10050874 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0265342
View Full Product Details