Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dpr6
Entrez Gene ID
50296
Gene Name
defective proboscis extension response 6
Gene Aliases
BcDNA:LD13525, CG14162, Dmel\CG14162, Dmel_CG14162, Dpr-6, Dpr6
Chromosome Location
Chr.3L: 9974651 - 10139360 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040823
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274744.2 NP_001261673.1 2 - 3 1716 63
NM_001274745.1 NP_001261674.1 2 - 3 1716 63
NM_001274746.1 NP_001261675.1 2 - 3 1716 63
NM_001300089.2 NP_001287018.1 2 - 3 1716 63
NM_144182.2 NP_652439.1 2 - 3 1716 63
GenBank mRNA AY069421.1 - 2 - 3 1748 63

View Full Product Details