Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Asx
Entrez Gene ID
36612
Gene Name
Additional sex combs
Gene Aliases
ASX, CG8787, Dmel\CG8787, Dmel_CG8787, FBgn0261823, xen
Chromosome Location
Chr.2R: 14503958 - 14512493 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261823
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014527.2 NP_001014527.1 8 - 9 4584 55
NM_001274040.1 NP_001260969.1 9 - 10 4647 55
NM_001274041.1 NP_001260970.1 8 - 9 5048 55
NM_166057.2 NP_725398.1 9 - 10 4913 55

View Full Product Details