Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
row
Entrez Gene ID
36685
Gene Name
relative of woc
Gene Aliases
CG8092, Dmel\CG8092, Dmel_CG8092, Row, anon-WO0118547.194
Chromosome Location
Chr.2R: 15206527 - 15213634 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033998
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274063.2 NP_001260992.1 1 - 2 1029 68
NM_137175.4 NP_611019.2 1 - 2 1029 68
NM_166081.3 NP_725441.1 1 - 2 1029 68
GenBank mRNA AY061301.1 - 1 - 2 998 68
BT016036.1 - 1 - 2 1008 68

View Full Product Details