Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncc69
Entrez Gene ID
39410
Gene Name
sodium chloride cotransporter 69
Gene Aliases
BEST:CK01027, CG43567, CG4357, CK01027, Dmel\CG4357, Dmel_CG4357, NCC69, NKCC, mdcds_25302
Chromosome Location
Chr.3L: 12368646 - 12391444 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036279
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274827.1 NP_001261756.1 6 - 7 2259 66
NM_001274828.1 NP_001261757.1 6 - 7 1433 66
NM_001274829.1 NP_001261758.1 6 - 7 1433 66
NM_001274830.2 NP_001261759.1 6 - 7 1488 66
NM_140315.4 NP_648572.1 6 - 7 1433 66
NM_206344.2 NP_996066.1 6 - 7 2259 66
GenBank mRNA AY094735.1 - 6 - 7 1404 66

View Full Product Details