The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272524.1 | NP_001259453.1 | 13 - 14 | 5153 | 55 |
NM_167292.3 | NP_727544.2 | 11 - 12 | 5087 | 55 | |
NM_206690.2 | NP_996413.2 | 11 - 12 | 5164 | 55 | |
NM_206691.3 | NP_996414.2 | 11 - 12 | 5336 | 55 |