Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptp10D
Entrez Gene ID
32115
Gene Name
Protein tyrosine phosphatase 10D
Gene Aliases
10D, CG1817, CT4920, DPTP, DPTP10D, DPTP[[10D]], Dmel\CG1817, Dmel_CG1817, PTP10D, Ptp10, R-PTP 10D, RPTP10d, dptp, ptp10D, ptp10d
Chromosome Location
Chr.X: 11622015 - 11677338 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004370
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_167292.3 NP_727544.2 12 - 13 6383 59
NM_206690.2 NP_996413.2 12 - 13 6460 59
NM_206691.3 NP_996414.2 12 - 13 6632 59

View Full Product Details