Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptp10D
Entrez Gene ID
32115
Gene Name
Protein tyrosine phosphatase 10D
Gene Aliases
10D, CG1817, CT4920, DPTP, DPTP10D, DPTP[[10D]], Dmel\CG1817, Dmel_CG1817, PTP10D, Ptp10, R-PTP 10D, RPTP10d, dptp, ptp10D, ptp10d
Chromosome Location
Chr.X: 11622015 - 11677338 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004370
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144715.2 NP_001138187.1 9 - 10 4877 67
NM_001272524.1 NP_001259453.1 11 - 12 4771 67
NM_001272525.1 NP_001259454.1 10 - 11 4750 67
NM_167292.3 NP_727544.2 9 - 10 4705 67
NM_206690.2 NP_996413.2 9 - 10 4782 67
NM_206691.3 NP_996414.2 9 - 10 4954 67

View Full Product Details