Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vap
Entrez Gene ID
32569
Gene Name
vacuolar peduncle
Gene Aliases
CG9209, D-RasGAP, Dmel\CG9209, Dmel_CG9209, FBpp0073946, RAS-GAP, RasGAP, RasGAP/Vap, RasGap, p120 RasGAP, p120RasGAP, rasGap
Chromosome Location
Chr.X: 15999951 - 16005812 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003969
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298385.1 NP_001285314.1 8 - 9 3704 89
NM_001298386.1 NP_001285315.1 9 - 10 3486 89
NM_078637.3 NP_523361.1 8 - 9 3067 89
NM_167480.2 NP_727925.1 9 - 10 3184 89
NM_167481.3 NP_727926.2 8 - 9 3369 89
NM_206759.2 NP_996482.1 10 - 11 3290 89
GenBank mRNA AJ012609.1 - 8 - 9 3006 89
BT044449.1 - 8 - 9 3067 89

View Full Product Details