Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rcp
Entrez Gene ID
32678
Gene Name
Receptor component protein
Gene Aliases
CG4875, Dmel\CG4875, Dmel_CG4875, dRCP
Chromosome Location
Chr.X: 16884581 - 16886612 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030801
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132947.5 NP_573175.2 2 - 3 148 73
NM_206770.2 NP_996493.1 2 - 3 148 73

View Full Product Details