Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ir20a
- Entrez Gene ID
-
33126
- Gene Name
- Ionotropic receptor 20a
- Gene Aliases
-
CG14584, DmelIR20a, Dmel\CG14584, Dmel_CG14584, IR20a
- Chromosome Location
-
Chr.X: 21477254 - 21478945 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031181
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134612.1 |
NP_608456.1 |
1 - 1 |
1614 |
57 |
View Full Product Details