Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfb4
Entrez Gene ID
33294
Gene Name
Transcription factor B4
Gene Aliases
CG5041, Dmel\CG5041, Dmel_CG5041, Dmp34, TFB4, p34
Chromosome Location
Chr.2L: 1149445 - 1150677 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031309
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134730.3 NP_608574.2 2 - 3 244 66
GenBank mRNA BT025234.2 - 2 - 3 240 66

View Full Product Details