Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eys
Entrez Gene ID
3771890
Gene Name
eyes shut
Gene Aliases
21A6, CG15388, CG15389, CG33955, CG7245, CT35437, Dmel\CG33955, Dmel_CG33955, EYS, Eyes Shut, Eys, MAb21A6, SPAM, Spam, mAb 21A6, mAb21A6, spacemaker, spam
Chromosome Location
Chr.2L: 2311680 - 2358944 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031414
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032399.3 NP_001027571.3 10 - 11 5751 70
NM_001272995.1 NP_001259924.2 10 - 11 5751 70
GenBank mRNA DQ991915.1 - 10 - 11 5738 70

View Full Product Details